Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phenylarsine Oxide

Catalog No. 521000250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-100-0250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-100-0250MG Supplier MilliporeSigma™ Supplier No. 521000250MG
Add to Cart
Add to Cart

A membrane-permeable protein tyrosine phosphatase inhibitor (IC50 = 18 μM).

A membrane-permeable protein tyrosine phosphatase inhibitor (IC50 = 18μM). Stimulates 2-deoxyglucose transport in insulin-resistant human skeletal muscle and activates p56 lck protein tyrosine kinase. Blocks TNF-α-dependent activation of NF-κB in human myeloid ML-1a cells. PAO inhibits the protease activities of recombinant human caspases as well as endogenous caspases that are active in extracts of pre-apoptotic chicken DU249 cells (S/M extracts).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 96% by titration
Target Proteins Protein tyrosine phosphatase
CAS 637-03-6
Molecular Formula C6H5AsO
Formula Weight 168g/mol
Solubility CHCl3 or DMSO
Form Solid
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.