CAS RN 637-07-0
Clofibrate, 95+%
CAS: 637-07-0 Molecular Formula: C12H15ClO3 Molecular Weight (g/mol): 242.70 MDL Number: MFCD00000615 InChI Key: KNHUKKLJHYUCFP-UHFFFAOYSA-N Synonym: clofibrate,ethyl clofibrate,clofibratum,clofibrato,atromid-s,miscleron,atromid,liprin,epib,angiokapsul PubChem CID: 2796 ChEBI: CHEBI:3750 IUPAC Name: ethyl 2-(4-chlorophenoxy)-2-methylpropanoate SMILES: CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
Clofibrate 98.0+%, TCI America™
CAS: 637-07-0 Molecular Formula: C12H15ClO3 Molecular Weight (g/mol): 242.70 MDL Number: MFCD00000615 InChI Key: KNHUKKLJHYUCFP-UHFFFAOYSA-N Synonym: clofibrate,ethyl clofibrate,clofibratum,clofibrato,atromid-s,miscleron,atromid,liprin,epib,angiokapsul PubChem CID: 2796 ChEBI: CHEBI:3750 IUPAC Name: ethyl 2-(4-chlorophenoxy)-2-methylpropanoate SMILES: CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
Clofibrate, 98%, MP Biomedicals™
CAS: 637-07-0 Molecular Formula: C12H15ClO3 Molecular Weight (g/mol): 242.70 MDL Number: MFCD00000615 InChI Key: KNHUKKLJHYUCFP-UHFFFAOYSA-N Synonym: clofibrate,ethyl clofibrate,clofibratum,clofibrato,atromid-s,miscleron,atromid,liprin,epib,angiokapsul PubChem CID: 2796 ChEBI: CHEBI:3750 IUPAC Name: ethyl 2-(4-chlorophenoxy)-2-methylpropanoate SMILES: CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
Clofibrate, R&D Systems™
CAS: 637-07-0 Molecular Formula: C12H15ClO3 Molecular Weight (g/mol): 242.70 MDL Number: MFCD00000615 InChI Key: KNHUKKLJHYUCFP-UHFFFAOYSA-N Synonym: clofibrate,ethyl clofibrate,clofibratum,clofibrato,atromid-s,miscleron,atromid,liprin,epib,angiokapsul PubChem CID: 2796 ChEBI: CHEBI:3750 IUPAC Name: ethyl 2-(4-chlorophenoxy)-2-methylpropanoate SMILES: CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1