accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 637-21-8

IUPAC Name: hydrogen 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-hydroxy-2-phenylacetate chloride
Synonyms: hydrogen homatropine chloride
Molecular Weight (g/mol): 311.81
Molecular Formula: C16H22ClNO3
InChi Key: FRMDDIJBLQNNTC-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN1C2CCC1CC(C2)OC(=O)C(O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results