Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homatropine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. H015625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H015625G 25 g
H01561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H015625G Supplier TCI America Supplier No. H015625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 637-21-8
Molecular Formula C16H22ClNO3
Molecular Weight (g/mol) 311.806
MDL Number MFCD00050613
InChI Key FRMDDIJBLQNNTC-ISXUQMITSA-N
PubChem CID 76966341
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;hydrochloride
SMILES CN1C2CCC1CC(C2)OC(=O)C(C3=CC=CC=C3)O.Cl

Specifications

Melting Point 220°C
Color White
Quantity 25 g
UN Number 1544
Formula Weight 311.81
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Homatropine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.