accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3ONH2

CAS RN 637-56-9

IUPAC Name: hydrogen 4-ethoxyaniline chloride
Synonyms: hydrogen P-ethoxyaniline chloride
Molecular Weight (g/mol): 173.64
Molecular Formula: C8H12ClNO
InChi Key: JVWYCUBGJVOEIZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC1=CC=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results