accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COONO

CAS RN 637-58-1

IUPAC Name: 4-[3-(4-butoxyphenoxy)propyl]morpholine hydrochloride
Synonyms: pramoxine hydrochloride
Molecular Weight (g/mol): 329.87
Molecular Formula: C17H28ClNO3
InChi Key: SYCBXBCPLUFJID-UHFFFAOYSA-N
SMILES: Cl.CCCCOC1=CC=C(OCCCN2CCOCC2)C=C1
Grade 
  • (1)
  • (7)
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (1)
  • (6)
Physical Form 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (7)
  • (1)
  • (1)
  • (6)
  • (6)
  • (1)
  • (7)
  • (1)
  • (6)
  • (1)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)