Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Pramoxine Hydrochloride USP
SDP

Catalog No. 14185554 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-554 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-554 Supplier Medisca Supplier No. 045804
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pramoxine Hydrochloride USP is a white to practically white, crystalline powder. May have a slight aromatic odor. Alternative name(s): Pramocaine Hydrochloride

Pramoxine Hydrochloride USP is a white to practically white, crystalline powder. May have a slight aromatic odor.

Alternative name(s): Pramocaine Hydrochloride

Specifications

Chemical Name or Material Pramoxine Hydrochloride USP
Packaging 120cc Amber Glass Round w/m bottle
CAS 637-58-1
Type Drug Standard
Molecular Weight (g/mol) 329.8652
Quantity 25 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.