accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3CH3H3C

CAS RN 637-92-3

IUPAC Name: 2-ethoxy-2-methylpropane
Synonyms: ethyl tert-butyl ether ethyl T-butyl ether
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: NUMQCACRALPSHD-UHFFFAOYSA-N
SMILES: CCOC(C)(C)C
Molecular Weight (g/mol) 
  • (4)
  • (8)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (5)
  • (4)
  • (8)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)