CAS RN 63721-05-1
IUPAC Name:
methyl 3,3-dimethylpent-4-enoate
Synonyms:
methyl 3,3-dimethylpent-4-enoate
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
MKLKDUHMZCIBSJ-UHFFFAOYSA-N
SMILES:
COC(=O)CC(C)(C)C=C
1
–
1
of
1
results
1
Methyl 3,3-Dimethyl-4-pentenoate 98.0+%, TCI America™
CAS: 63721-05-1 Molecular Formula: C8H14O2 Molecular Weight (g/mol): 142.198 MDL Number: MFCD00075381 InChI Key: MKLKDUHMZCIBSJ-UHFFFAOYSA-N Synonym: 3,3-Dimethyl-4-pentenoic Acid Methyl Ester PubChem CID: 114129 IUPAC Name: methyl 3,3-dimethylpent-4-enoate SMILES: CC(C)(CC(=O)OC)C=C