Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,3-Dimethyl-4-pentenoate 98.0+%, TCI America™
SDP

Catalog No. D215325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D215325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D215325ML Supplier TCI America Supplier No. D215325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63721-05-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00075381
InChI Key MKLKDUHMZCIBSJ-UHFFFAOYSA-N
Synonym 3,3-Dimethyl-4-pentenoic Acid Methyl Ester
PubChem CID 114129
IUPAC Name methyl 3,3-dimethylpent-4-enoate
SMILES CC(C)(CC(=O)OC)C=C

Specifications

Color Colorless
Boiling Point 59°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3,3-Dimethyl-4-pentenoate
EINECSNumber (2)-3535
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.