accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 638-04-0

IUPAC Name: (1R,3S)-1,3-dimethylcyclohexane
Synonyms: (1R,3S)-1,3-dimethylcyclohexane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: SGVUHPSBDNVHKL-OCAPTIKFSA-N
SMILES: C[C@H]1CCC[C@@H](C)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results