Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,3-Dimethylcyclohexane 99.0+%, TCI America™
SDP

Catalog No. D10555ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10555ML 5 mL
D105525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10555ML Supplier TCI America Supplier No. D10555ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 638-04-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00001507
InChI Key SGVUHPSBDNVHKL-OCAPTIKFSA-N
Synonym cis-1,3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexane, cyclohexane, 1,3-dimethyl-, cis, 1,cis-3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexan, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, cis-hexahydro-m-xylene, c-1,3-dimethylcyclohexane, 1,3-dimethyl cis-cyclohexane
PubChem CID 252361
IUPAC Name (1R,3S)-1,3-dimethylcyclohexane
SMILES C[C@H]1CCC[C@@H](C)C1

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 mL
UN Number 2263
Formula Weight 112.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-1,3-Dimethylcyclohexane
EINECSNumber (3)-2227
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.