CAS RN 638-67-5
Tricosane, 98%
CAS: 638-67-5 Molecular Formula: C23H48 Molecular Weight (g/mol): 324.63 MDL Number: MFCD00009350 InChI Key: FIGVVZUWCLSUEI-UHFFFAOYSA-N Synonym: n-tricosane,tricosan,docosane, methyl,unii-t166b8r1vc,ch3-ch2 21-ch3,tricosane, analytical standard,tricosane,tricosane standardmaterialforgc,unii-j3n6x3yk96 component PubChem CID: 12534 ChEBI: CHEBI:32934 IUPAC Name: tricosane SMILES: CCCCCCCCCCCCCCCCCCCCCCC
n-Tricosane, 99%
CAS: 638-67-5 Molecular Formula: C23H48 Molecular Weight (g/mol): 324.637 MDL Number: MFCD00009350 InChI Key: FIGVVZUWCLSUEI-UHFFFAOYSA-N Synonym: n-tricosane,tricosan,docosane, methyl,unii-t166b8r1vc,ch3-ch2 21-ch3,tricosane, analytical standard,tricosane,tricosane standardmaterialforgc,unii-j3n6x3yk96 component PubChem CID: 12534 ChEBI: CHEBI:32934 IUPAC Name: tricosane SMILES: CCCCCCCCCCCCCCCCCCCCCCC
Tricosane 98.0+%, TCI America™
CAS: 638-67-5 Molecular Formula: C23H48 Molecular Weight (g/mol): 324.637 MDL Number: MFCD00009350 InChI Key: FIGVVZUWCLSUEI-UHFFFAOYSA-N Synonym: n-tricosane,tricosan,docosane, methyl,unii-t166b8r1vc,ch3-ch2 21-ch3,tricosane, analytical standard,tricosane,tricosane standardmaterialforgc,unii-j3n6x3yk96 component PubChem CID: 12534 ChEBI: CHEBI:32934 IUPAC Name: tricosane SMILES: CCCCCCCCCCCCCCCCCCCCCCC
Tricosane 99.0+%, TCI America™
CAS: 638-67-5 Molecular Formula: C23H48 Molecular Weight (g/mol): 324.637 MDL Number: MFCD00009350 InChI Key: FIGVVZUWCLSUEI-UHFFFAOYSA-N Synonym: n-tricosane,tricosan,docosane, methyl,unii-t166b8r1vc,ch3-ch2 21-ch3,tricosane, analytical standard,tricosane,tricosane standardmaterialforgc,unii-j3n6x3yk96 component PubChem CID: 12534 ChEBI: CHEBI:32934 IUPAC Name: tricosane SMILES: CCCCCCCCCCCCCCCCCCCCCCC
Sigma Aldrich Tricosane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc n-Tricosane, 97%, MP Biomedicals
CAS: 638-67-5 Molecular Formula: C23H48 Molecular Weight (g/mol): 324.637 InChI Key: FIGVVZUWCLSUEI-UHFFFAOYSA-N Synonym: n-tricosane,tricosan,docosane, methyl,unii-t166b8r1vc,ch3-ch2 21-ch3,tricosane, analytical standard,tricosane,tricosane standardmaterialforgc,unii-j3n6x3yk96 component PubChem CID: 12534 ChEBI: CHEBI:32934 IUPAC Name: tricosane SMILES: CCCCCCCCCCCCCCCCCCCCCCC
n-Tricosane, SPEX CertiPrep™
CAS: 638-67-5 Molecular Formula: C23H48 Molecular Weight (g/mol): 324.637 InChI Key: FIGVVZUWCLSUEI-UHFFFAOYSA-N PubChem CID: 12534 ChEBI: CHEBI:32934 IUPAC Name: tricosane SMILES: CCCCCCCCCCCCCCCCCCCCCCC