accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(R)H3C(S)CH3(R)(R)H3C(S)OHCH3H3C(R)(R)CH3(R)(S)CH3

CAS RN 638-95-9

IUPAC Name: (3S,4aR,6aR,6bS,8aR,11R,12S,12aR,14aR,14bR)-4,4,6a,6b,8a,11,12,14b-octamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-ol
Synonyms: α-amyrin α-amyrine viminalol α-amyrenol urs-12-ene-3β-ol
Molecular Weight (g/mol): 426.73
Molecular Formula: C30H50O
InChi Key: FSLPMRQHCOLESF-SFMCKYFRSA-N
SMILES: C[C@@H]1CC[C@]2(C)CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results