Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Amyrin, MP Biomedicals

Catalog No. ICN220236 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN220236 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN220236 Supplier MP Biomedicals, Inc Supplier No. 0522023610

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 638-95-9
Molecular Formula C30H50O
Molecular Weight (g/mol) 426.729
InChI Key FSLPMRQHCOLESF-SFMCKYFRSA-N
Synonym alpha-amyrin, viminalol, alpha-amyrenol, alpha-amyrine, urs-12-en-3beta-ol, unii-30zag40j8n, 5alpha-urs-12-en-3beta-ol, 3beta-urs-12-en-3-ol, a-amyrin, 3s,4ar,6ar,6bs,8ar,11r,12s,12ar,14ar,14br-4,4,6a,6b,8a,11,12,14b-octamethyl-2,3,4a,5,6,7,8,9,10,11,12,12a,14,14a-tetradecahydro-1h-picen-3-ol
PubChem CID 73170
ChEBI CHEBI:10213
IUPAC Name (3S,4aR,6aR,6bS,8aR,11R,12S,12aR,14aR,14bR)-4,4,6a,6b,8a,11,12,14b-octamethyl-2,3,4a,5,6,7,8,9,10,11,12,12a,14,14a-tetradecahydro-1H-picen-3-ol
SMILES CC1CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1C)C)C

Specifications

Quantity 100 mg
Formula Weight 426.39
Chemical Name or Material α-Amyrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.