accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONHCl

CAS RN 6380-05-8

IUPAC Name: N-(2-chlorophenyl)-4-methylbenzene-1-sulfonamide
Synonyms: N-(2-chlorophenyl)-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 281.75
Molecular Formula: C13H12ClNO2S
InChi Key: GOEWZPICEGNTPR-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results