Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aceanthrenequinone 99+%, Thermo Scientific™

Catalog No. AC292790010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC292790010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292790010 Supplier Thermo Scientific Chemicals Supplier No. 292790010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6380-05-8
Molecular Formula C13H12ClNO2S
Molecular Weight (g/mol) 281.75
InChI Key GOEWZPICEGNTPR-UHFFFAOYSA-N
Synonym n-2-chlorophenyl-4-methylbenzenesulfonamide, n-2-chlorophenyl-p-toluenesulphonamide, n-tosyl-2-chloroaniline, n-2-chlorophenyl-p-toluenesulfonamide, 2-chlorophenyl 4-methylphenyl sulfonyl amine, n-2-chloro-phenyl-4-methyl-benzenesulfonamide, benzenesulfonamide, n-2-chlorophenyl-4-methyl, n-2-chlorophenyl-4-methylbenzene-1-sulfonamide, n-2-chlorophenyl-4-methyl-benzenesulfonamide
PubChem CID 80791
IUPAC Name N-(2-chlorophenyl)-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 268°C to 271°C
Color Orange to Brown
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic
Quantity 1 g
Beilstein 07, 436
Formula Weight 232.24
Physical Form Powder
Chemical Name or Material Aceanthrenequinone, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.