accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNBrClN

CAS RN 63810-78-6

IUPAC Name: 5-bromo-4-chloro-2-(methylsulfanyl)pyrimidine
Synonyms: 5-bromo-4-chloro-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 239.52
Molecular Formula: C5H4BrClN2S
InChi Key: XVRYJQACDDVZEI-UHFFFAOYSA-N
SMILES: CSC1=NC=C(Br)C(Cl)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results