accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSBrN

CAS RN 63837-11-6

IUPAC Name: 5-bromo-2-methyl-1,3-benzothiazole
Synonyms: 5-bromo-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol): 228.11
Molecular Formula: C8H6BrNS
InChi Key: OLQKNZNXLBILDD-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(Br)=CC=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)