Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methylbenzothiazole 98.0+%, TCI America™
SDP

Catalog No. B17985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17985G Supplier TCI America Supplier No. B17985G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

CAS 63837-11-6
Melting Point 79°C
Color White-Yellow
Boiling Point 170°C
Molecular Formula C8H6BrNS
Quantity 5 g
MDL Number MFCD00603096
InChI Key OLQKNZNXLBILDD-UHFFFAOYSA-N
SMILES CC1=NC2=C(S1)C=CC(=C2)Br
IUPAC Name 5-bromo-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol) 228.107
PubChem CID 3017457
Formula Weight 228.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methylbenzothiazole
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.