accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63922-84-9

IUPAC Name: triphenyl[2-(trimethylsilyl)ethyl]phosphanium iodide
Synonyms: triphenyl[2-(trimethylsilyl)ethyl]phosphanium iodide
Molecular Weight (g/mol): 490.44
Molecular Formula: C23H28IPSi
InChi Key: NXZYPOSEOPBJMW-UHFFFAOYSA-M
SMILES: [I-].C[Si](C)(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results