Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Trimethylsilylethyl)triphenylphosphonium Iodide 98.0+%, TCI America™
SDP

Catalog No. T15105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15105G 5 g
T15101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15105G Supplier TCI America Supplier No. T15105G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63922-84-9
Molecular Formula C23H28IPSi
Molecular Weight (g/mol) 490.44
MDL Number MFCD00191591
InChI Key NXZYPOSEOPBJMW-UHFFFAOYSA-M
Synonym Triphenyl(2-trimethylsilylethyl)phosphonium Iodide
PubChem CID 22245266
IUPAC Name triphenyl(2-trimethylsilylethyl)phosphanium;iodide
SMILES C[Si](C)(C)CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 5 g
Formula Weight 490.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Trimethylsilylethyl)triphenylphosphonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.