accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSFF

CAS RN 63930-17-6

IUPAC Name: 1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene
Synonyms: 1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene
Molecular Weight (g/mol): 254.31
Molecular Formula: C12H8F2S2
InChi Key: LFVFHTBHAIVNJQ-UHFFFAOYSA-N
SMILES: FC1=CC(SSC2=CC=CC(F)=C2)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results