Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3-fluorophenyl) Disulfide 97.0+%, TCI America™
SDP

Catalog No. B30445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30445G 5 g
B30441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30445G Supplier TCI America Supplier No. B30445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63930-17-6
Molecular Formula C12H8F2S2
Molecular Weight (g/mol) 254.313
MDL Number MFCD01318104
InChI Key LFVFHTBHAIVNJQ-UHFFFAOYSA-N
Synonym bis 3-fluorophenyl disulfide, bis 3-fluorophenyl disulphide, 1,2-bis 3-fluorophenyl disulfane, 3,3'-difluoro diphenyl disulfide, 1-fluoro-3-3-fluorophenyl disulfanyl benzene, 3,3'-difluorodiphenyl disulfide, bis-3-fluorophenyl disulfide, disulfide, bis 3-fluorophenyl, di3-fluorophenyl disulfide
PubChem CID 2737794
IUPAC Name 1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene
SMILES C1=CC(=CC(=C1)SSC2=CC=CC(=C2)F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 254.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(3-fluorophenyl) Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.