accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63939-65-1

IUPAC Name: trimethyl[4-(2-oxopyrrolidin-1-yl)but-2-yn-1-yl]azanium
Synonyms: oxotremorine M oxotremorine-M
Molecular Weight (g/mol): 195.29
Molecular Formula: C11H19N2O
InChi Key: CANZROMYQDHYHR-UHFFFAOYSA-N
SMILES: C[N+](C)(C)CC#CCN1CCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results