Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orcinol, 99.6%, MP Biomedicals™

Catalog No. ICN10250650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10250650 50 g
ICN10250605 5 g
ICN10250610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10250650 Supplier MP Biomedicals, Inc Supplier No. 0210250650
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63939-65-1
Molecular Formula C11H19IN2O
Molecular Weight (g/mol) 322.19
InChI Key VVLMSCJCXMBGDI-UHFFFAOYSA-M
Synonym oxotremorine m, oxotremorine-m, 3h oxotremorine-m, n,n,n-trimethyl-4-2-oxopyrrolidin-1-yl but-2-yn-1-aminium, trimethyl-4-2-oxopyrrolidin-1-yl but-2-ynyl azanium, n,n,n-trimethyl-4-2-oxo-1-pyrrolidinyl-2-butyn-1-aminium, 2-butyn-1-aminium, n,n,n-trimethyl-4-2-oxo-1-pyrrolidinyl, chembl23957, spectrum_001982, tocris-1067
PubChem CID 3027782
IUPAC Name trimethyl-[4-(2-oxopyrrolidin-1-yl)but-2-ynyl]azanium;iodide
SMILES C[N+](C)(C)CC#CCN1CCCC1=O.[I-]

Specifications

Color Tan to White
Assay Percent Range ∽ 98%
Quantity 50 g
Formula Weight 142.2
Percent Purity 99.6%
Physical Form Powder
Chemical Name or Material Orcinol
EINECSNumber 207-984-2
storageNote1 Store Desiccated.. Protect from light
Recommended Storage Store at room temperature (15° to 30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.