accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63968-84-3

IUPAC Name: N'-hydroxy-2-(trifluoromethoxy)benzene-1-carboximidamide
Synonyms: N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
Molecular Weight (g/mol): 220.15
Molecular Formula: C8H7F3N2O2
InChi Key: KGHGHEPVWUOJAZ-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC=CC=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results