Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N'-Hydroxy-2-(trifluoromethoxy)benzenecarboximidamide, 95%, Thermo Scientific™

Catalog No. MO01204EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01204EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01204EA Supplier Thermo Scientific Chemicals Supplier No. MO01204EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63968-84-3
Molecular Formula C8H7F3N2O2
Molecular Weight (g/mol) 220.15
MDL Number MFCD07772878
InChI Key KGHGHEPVWUOJAZ-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzamidoxime, ho dpmlgbprprprjpjjiqy jhhfhabp, n'-hydroxy-2-trifluoromethoxy benzimidamide, hydroxyimino 2-trifluoromethoxy phenyl methylamine, benzenecarboximidamide, n-hydroxy-2-trifluoromethoxy, z-n'-hydroxy-2-trifluoromethoxy benzene-1-carboximidamide
PubChem CID 9582829
SMILES NC(=NO)C1=CC=CC=C1OC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
Formula Weight 220.15
Percent Purity 95%
Chemical Name or Material N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.