accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 63975-59-7

IUPAC Name: 4a-methyl-2,3,4,4a,5,6,7,8-octahydronaphthalen-2-one
Synonyms: 4a-methyl-3,4,5,6,7,8-hexahydronaphthalen-2-one
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: OHERZLWVBJCXOF-UHFFFAOYNA-N
SMILES: CC12CCCCC1=CC(=O)CC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results