CAS RN 6398-98-7
CAS RN 6398-98-7
4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 98%
CAS: 6398-98-7 Molecular Formula: C20H28Cl3N3O3 Molecular Weight (g/mol): 464.812 MDL Number: MFCD00078857 InChI Key: YVNAYSHNIILOJS-UHFFFAOYSA-N Synonym: amodiaquin dihydrochloride dihydrate,amodiaquine hcl,amodiaquine hydrochloride,amodiaquine dihydrochloride dihydrate,unii-k6pw2s574l,4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate,amodiaquine hydrochloride usp,cam-aq 1 PubChem CID: 64646 ChEBI: CHEBI:50652 IUPAC Name: 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride SMILES: CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl
Amodiaquin Dihydrochloride Dihydrate 98.0+%, TCI America™
CAS: 6398-98-7 Molecular Formula: C20H28Cl3N3O3 Molecular Weight (g/mol): 464.812 MDL Number: MFCD00078857 InChI Key: YVNAYSHNIILOJS-UHFFFAOYSA-N Synonym: amodiaquin dihydrochloride dihydrate,amodiaquine hcl,amodiaquine hydrochloride,amodiaquine dihydrochloride dihydrate,unii-k6pw2s574l,4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate,amodiaquine hydrochloride usp,cam-aq 1 PubChem CID: 64646 ChEBI: CHEBI:50652 IUPAC Name: 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride SMILES: CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl
Sigma Aldrich Amodiaquin dihydrochloride dihydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Thermo Scientific Chemicals Amodiaquine dihydrochloride dihydrate
CAS: 6398-98-7 Molecular Formula: 2HCl·2H2O Molecular Weight (g/mol): 464.81 InChI Key: YVNAYSHNIILOJS-UHFFFAOYSA-N Synonym: amodiaquin dihydrochloride dihydrate,amodiaquine hcl,amodiaquine hydrochloride,amodiaquine dihydrochloride dihydrate,unii-k6pw2s574l,4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate,amodiaquine hydrochloride usp,cam-aq 1 PubChem CID: 64646 ChEBI: CHEBI:50652 IUPAC Name: 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride SMILES: CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl