Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amodiaquin dihydrochloride dihydrate, MilliporeSigma™ Supelco™

Catalog No. 111015034 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5034 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5034 Supplier MilliporeSigma Supelco Supplier No. A27995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 6398-98-7
Physical Form Neat
Linear Formula C20H22 ClN3 O · 2 HCl · 2H2 O
MDL Number MFCD00078857
Synonym 4-([7-Chloro-4-quinolinyl]amino)-2-([diethylamino]methyl)phenol
Type Analytical Standard
Quantity 5 g
Formula Weight 464.81
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.