accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63989-40-2

IUPAC Name: 2-[(2-hydroxyethyl)amino]-5-nitrobenzonitrile
Synonyms: 2-[(2-hydroxyethyl)amino]-5-nitrobenzonitrile
Molecular Weight (g/mol): 207.19
Molecular Formula: C9H9N3O3
InChi Key: AADOGXABUHCROC-UHFFFAOYSA-N
SMILES: OCCNC1=C(C=C(C=C1)[N+]([O-])=O)C#N
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results