Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline, TCI America™
SDP

Catalog No. H07495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07495G Supplier TCI America Supplier No. H07495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63989-40-2
Molecular Formula C9H9N3O3
Molecular Weight (g/mol) 207.189
MDL Number MFCD00051934
InChI Key AADOGXABUHCROC-UHFFFAOYSA-N
Synonym 2-Cyano-N-(2-hydroxyethyl)-4-nitroaniline, 2-(2-Hydroxyethylamino)-5-nitrobenzonitrile
PubChem CID 600590
IUPAC Name 2-(2-hydroxyethylamino)-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)NCCO

Specifications

Melting Point 163°C
Color Yellow
Quantity 5 g
Formula Weight 207.19
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.