CAS RN 63995-70-0
IUPAC Name:
trisodium 3-[bis(3-sulfonatophenyl)phosphanyl]benzene-1-sulfonate
Synonyms:
tppts
Molecular Weight (g/mol):
568.41
Molecular Formula:
C18H12Na3O9PS3
InChi Key:
MYAJTCUQMQREFZ-UHFFFAOYSA-K
SMILES:
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)P(C1=CC(=CC=C1)S([O-])(=O)=O)C1=CC(=CC=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Sigma Aldrich Triphenylphosphine-3,3',3-Trisulfonic Acid Trisodium Salt
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.