accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64-04-0

IUPAC Name: 2-phenylethan-1-amine
Synonyms: 2-phenylethylamine β-phenethylamine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: BHHGXPLMPWCGHP-UHFFFAOYSA-N
SMILES: NCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (9)
  • (7)
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (3)
  • (8)
  • (9)
  • (7)
  • (3)
  • (1)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (8)