Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenethylamine

Catalog No. 407267800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-726-7800ML 800 mL
40-726-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-726-7800ML Supplier Sigma Aldrich Supplier No. 407267800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phenethylamine (α-phenylethylamine, pea) is a biogenic amine
  • Its structure and properties are similar to amphetamine
  • The influence of pea on dopamine concentrations in the nucleus accumbens and prefrontal cortex has been investigated in rats
  • N-alkylation of phenethylamine, employing alcohols as the alkylating agent has been reported
  • Normal raman (nr) and the surface-enhanced raman scattering (sers) for the characterization of phenethylamine has been reported.phenethylamine is the suitable reagent used for the preparation of bis(2-phenylethylammonium) tetrachloridocobaltate(ii)
  • It may be used for the conversion of c14o2 into a soluble form for liquid scintillation counting
  • It may be used for the synthesis of series of the layered inorganic-organic perovskite materials

Specifications

CAS 64-04-0
Density 0.962 g/mL (at 20°C (literature))
Boiling Point 197°C to 200°C (lit.)
Molecular Formula C8H11N
Refractive Index n20/D 1.533 (literature)
Linear Formula C6H5CH2CH2NH2
MDL Number MFCD00008184
Quantity 800 mL
Synonym 2-Phenethylamine; beta-Phenylethylamine; 2-Phenylethylamine
Molecular Weight (g/mol) 121.18
Percent Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.