accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 640-61-9

IUPAC Name: N,4-dimethylbenzene-1-sulfonamide
Synonyms: N,4-dimethylbenzenesulfonamide
Molecular Weight (g/mol): 185.24
Molecular Formula: C8H11NO2S
InChi Key: GWLOGZRVYXAHRE-UHFFFAOYSA-N
SMILES: CNS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results