Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-p-toluenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. M171725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M171725G 25 g
M1717250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M171725G Supplier TCI America Supplier No. M171725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 640-61-9
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
InChI Key GWLOGZRVYXAHRE-UHFFFAOYSA-N
Synonym n-methyl-p-toluenesulfonamide, benzenesulfonamide, n,4-dimethyl, p-tolylsulfonyl methylamine, p-toluene-n-methylsulfonamide, p-toluenesulfonamide, n-methyl, n,4-dimethyl-benzenesulfonamide, n-methyl-para-toluenesulfonamide, n,4-dimethylbenzene-1-sulfonamide, benzenesulfonamide,4-dimethyl, methyl 4-methylphenyl sulfonyl amine
PubChem CID 12543
IUPAC Name N,4-dimethylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 25 g
Formula Weight 185.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-p-toluenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.