CAS RN 64034-02-2
IUPAC Name:
heptadeca-2,4-diynoic acid
Synonyms:
heptadeca-2,4-diynoic acid
Molecular Weight (g/mol):
262.39
Molecular Formula:
C17H26O2
InChi Key:
JMRNAAJGQAFZRJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCC#CC#CC(O)=O
1
–
1
of
1
results
1
2,4-Heptadecadiynoic Acid 98.0+%, TCI America™
CAS: 64034-02-2 Molecular Formula: C17H26O2 Molecular Weight (g/mol): 262.393 MDL Number: MFCD00060064 InChI Key: JMRNAAJGQAFZRJ-UHFFFAOYSA-N PubChem CID: 24206697 IUPAC Name: heptadeca-2,4-diynoic acid SMILES: CCCCCCCCCCCCC#CC#CC(=O)O