Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Heptadecadiynoic Acid 98.0+%, TCI America™
SDP

Catalog No. H0667100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0667100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0667100MG Supplier TCI America Supplier No. H0667100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64034-02-2
Molecular Formula C17H26O2
Molecular Weight (g/mol) 262.393
MDL Number MFCD00060064
InChI Key JMRNAAJGQAFZRJ-UHFFFAOYSA-N
PubChem CID 24206697
IUPAC Name heptadeca-2,4-diynoic acid
SMILES CCCCCCCCCCCCC#CC#CC(=O)O

Specifications

Melting Point 60°C
Quantity 100 mg
Formula Weight 262.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Heptadecadiynoic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.