accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COSOOFFF

CAS RN 64035-64-9

IUPAC Name: (trimethylsilyl)methyl trifluoromethanesulfonate
Synonyms: (trimethylsilyl)methyl trifluoromethanesulfonate
Molecular Weight (g/mol): 236.28
Molecular Formula: C5H11F3O3SSi
InChi Key: VMDMAAJZSXXCQV-UHFFFAOYSA-N
SMILES: C[Si](C)(C)COS(=O)(=O)C(F)(F)F
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results