accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOOCH3CH2

CAS RN 64057-79-0

IUPAC Name: bis(2-methylprop-2-en-1-yl) carbonate
Synonyms: bis(2-methylprop-2-en-1-yl) carbonate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: SKCUYCAPQSWGBP-UHFFFAOYSA-N
SMILES: CC(=C)COC(=O)OCC(C)=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results