accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClZnH3CONNNOOOH3CNH2N(E/Z)

CAS RN 64071-86-9

IUPAC Name: zinc(2+) 2-{2,5-dimethoxy-4-[2-(4-nitrophenyl)diazen-1-yl]phenyl}diazenium trichloride
Synonyms: zinc(2+) 2-{2,5-dimethoxy-4-[2-(4-nitrophenyl)diazen-1-yl]phenyl}diazenium trichloride
Molecular Weight (g/mol): 488.03
Molecular Formula: C14H14Cl3N5O4Zn
InChi Key: KTSPHVBBBOGQDW-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Cl-].[Zn++].COC1=CC(N=NC2=CC=C(C=C2)[N+]([O-])=O)=C(OC)C=C1N=[NH2+]
Grade 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)