Learn More
Description
Fast black K (C. l. 37190) is a lipophilic amino-substituted monoazo dye. It is also known as Brentamine fast black K salt.
Specifications
Specifications
| Chemical Name or Material | Brentamine fast black K salt |
| CAS | 64071-86-9 |
| Color | Brown |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 417.88 g/mol |
| Grade | Practical |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.