accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64074-29-9

CH3OONN

CAS RN 64074-29-9

IUPAC Name: methyl 2-phenylpyrimidine-5-carboxylate
Synonyms: methyl 2-phenylpyrimidine-5-carboxylate
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: SOMJCOFUAUMQPG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results