accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaCH3NHNHH3CSOOOOOSOOO

CAS RN 6408-57-7

IUPAC Name: disodium 5-butyl-2-({4-[(4-butyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate
Synonyms: disodium 5-butyl-2-({4-[(4-butyl-2-sulfonatophenyl)amino]-9,10-dioxoanthracen-1-yl}amino)benzenesulfonate
Molecular Weight (g/mol): 706.74
Molecular Formula: C34H32N2Na2O8S2
InChi Key: BOXAUJCFZBSNKZ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CCCCC1=CC=C(NC2=CC=C(NC3=CC=C(CCCC)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results