Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Green 27 75.0+%, TCI America™
SDP

Catalog No. A186325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A186325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A186325G Supplier TCI America Supplier No. A186325G
Add to Cart
Add to Cart

Laboratory reagent with a purity of ≥75.0% (HPLC).

Chemical Identifiers

CAS 6408-57-7
Molecular Formula C34H32N2Na2O8S2
Molecular Weight (g/mol) 706.74
MDL Number MFCD00001196
InChI Key BOXAUJCFZBSNKZ-UHFFFAOYSA-L
PubChem CID 80837
IUPAC Name disodium 5-butyl-2-({4-[(4-butyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate
SMILES [Na+].[Na+].CCCCC1=CC=C(NC2=CC=C(NC3=CC=C(CCCC)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O

Specifications

Melting Point 260°C
Color Black
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 706.74
Percent Purity ≥75.0% (HPLC)
Grade Laboratory
Chemical Name or Material Acid Green 27
EINECSNumber (5)-1744
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.