accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64090-19-3

IUPAC Name: 1-(4-fluorophenyl)piperazine dihydrochloride
Synonyms: para-fluorophenylpiperazine dihydrochloride
Molecular Weight (g/mol): 253.14
Molecular Formula: C10H15Cl2FN2
InChi Key: DZQVAQAZQDURKX-UHFFFAOYSA-N
SMILES: Cl.Cl.FC1=CC=C(C=C1)N1CCNCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results