Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)piperazine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. F028825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F028825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F028825G Supplier TCI America Supplier No. F028825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64090-19-3
Molecular Formula C10H15Cl2FN2
Molecular Weight (g/mol) 253.142
MDL Number MFCD00012765
InChI Key DZQVAQAZQDURKX-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p
PubChem CID 16211916
IUPAC Name 1-(4-fluorophenyl)piperazine;dihydrochloride
SMILES C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl

Specifications

Quantity 25 g
Formula Weight 253.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Fluorophenyl)piperazine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.