CAS RN 6417-85-2
IUPAC Name:
dipotassium 3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Synonyms:
dipotassium 3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular Weight (g/mol):
408.49
Molecular Formula:
C20H10K2O5
InChi Key:
DZSPMLRDMGJVAT-UHFFFAOYSA-L
SMILES:
[K+].[K+].[O-]C1=CC=C2C(OC3=CC([O-])=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
1
–
1
of
1
results
1
Uranine K, TCI America™
CAS: 6417-85-2 Molecular Formula: C20H12O5 MDL Number: MFCD00068316 Synonym: Fluorescein Potassium Salt